missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ AloI, Fisher BioReagents™
Click to view available options
Quantity:
100 U
Unit Size:
Each
Description
5'...^ 7(N) G A A C (N)6 T C C (N)12-13^...3'
3'...^12-13(N) C T T G (N)6 A G G (N)7 ^...5'
Supplied with: 10X OPTIZYME Buffer 5
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
- Incubate at 30°C
- Buffer 1: 0 - 20%
- Buffer 2: 0 - 20%
- Buffer 3: 0 - 20%
- Buffer 4: 20 - 50%
- Buffer 5: 100%
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
- Incubation at 37°C results in 20% activity.
- AloI produces double-strand cuts on both sides from the interrupted recognition site. Its unique feature is a degenerate cleavage point on the 3 side of the recognition sequence (12 or 13 nucleotides away).
- The presence of S-adenosylmethionine in the reaction mixture results in incomplete cleavage with AloI.
- Greater than 10-fold overdigestion with AloI may result in star activity.
- AloI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.
Dam: May overlap - effect not determined
- 5'...GAAC(N)4Gm6A TCC...3'
- 5'...GAAC(N)6TCm5CW GG...3'
- 5'...m5C GAAC(N)6TCC...3'
- 5'...GAAC(N)6TCm5C G...3'
- 5'...GAAm5C G(N)5TCC...3'
Specifications
Specifications
| Concentration | 1 to 3 U/μL |
| Components | 25 to 50% Water, >50% Glycerin |
| Incubator Temperature | 30°C |
| pH | 7.4 |
| For Use With (Application) | >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with AloI |
| Storage Buffer | 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol |
| Content And Storage | Keep container tightly closed |
| Quantity | 100 U |
| Cut Site | (7/12)GAAC(N)6TCC(12/7) |
| Product Type | AloI |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction