missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ AarI, Fisher BioReagents™

Product Code. 11944822
Click to view available options
Quantity:
125 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11944822

Brand: Fisher BioReagents BP80691

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...C A C C T G C (N)4 ^...3'
3'...G T G G A C G (N)8 ^...5'

Supplied with: 10X OPTIZYME Buffer AarI, ScaI, PasI and 50X Oligonucleotide
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer AarI, ScaI, PasI: 10mM Bis-Tris Propane-HCl (pH 6.5 at 37°C), 10mM MgCl2, 100mM KCl, 0.1mg/ml BSA
  • 0.5μM of oligonucleotide
  • Incubate at 37°C
  • Enzyme Activity in OPTIZYME buffers:
    • Buffer 1: No Reaction (+Oligonucleotide)
  • Buffer 2: No Reaction (+Oligonucleotide)
  • Buffer 3: 0 - 20% (+Oligonucleotide)
  • Buffer 4: No Reaction (+Oligonucleotide)
  • Buffer 5: 0 - 20% (+Oligonucleotide)
  • Storage Buffer:
    • 10mM potassium phosphate (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 10-fold over-digestion with AarI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of AarI is required for the digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Notes:
    • At least two copies of its recognition sequence are required for cleavage with AarI
    • 0.5μM oligonucleotide improves cleavage of DNAs, especially those with a single AarI site.
    • Greater than 10-fold overdigestion with AarI may result in star activity.
    • AarI may remain associated with cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye and SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.

    Methylation Effects:
    Dam: Never overlaps - no effect
    Dcm: Never overlaps - no effect
    CpG: May overlap - cleavage impaired
    • 5'...CACCTGm5C G...3'
  • 3'...GTGGAC Gm5C...5'
  • (Cleavage is impaired)
  • EcoKI: Never overlaps - no effect
  • EcoBI: May overlap - effect not determined
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 2 U/μL
    Components 25 to 50% Water, >50% Glycerin
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 10-fold over-digestion with AarI
    Storage Buffer 10mM Potassium Phosphate (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 125 U
    Cut Site CACCTGC
    Product Type Aarl
    Form Liquid
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.