Learn More
Corning™ LSE™ Optional Head
Product Code. p-8009064 Shop All Corning Products
Click to view available options
Holds:
12 x 1.5 to 2 mL Tubes
2 x 50 mL Tubes
4 x 15 mL Tubes
Unit Size:
Each
Product Code. 11981981
Brand: Corning™ 480107
Only null left
Add to basket
Add to basket
For use with various sizes of tubes.
This is an optional head for tubes, held horizontally, for use with Corning™ LSE™ Vortex Mixer.
TRUSTED_SUSTAINABILITY
Specifications
Holds 2 x 50 mL Tubes
Product Type Optional Head
For Use With (Equipment) Corning™ LSE™ Vortex Mixer