missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Optineurin Antibody (S01-2C5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19900-100ul
This item is not returnable.
View return policy
Description
Optineurin Monoclonal antibody specifically detects Optineurin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Gene Knock-Out
Specifications
| Optineurin | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:50, Immunocytochemistry/ Immunofluorescence Validated for Immunocytochemistry/Immunofluorescence from YCharOS Inc. (ycharos.com)., Immunohistochemistry-Paraffin, Knockout Validated Validated for Knockout from YCharOS Inc. (ycharos.com). | |
| FIP2FIP-2, GLC1Eglaucoma 1, open angle, E (adult-onset), HIP7ALS12, Huntingtin interacting protein L, Huntingtin yeast partner L, Huntingtin-interacting protein 7, Huntingtin-interacting protein L, HYPLHIP-7, NEMO-related protein, NRPE3-14.7K-interacting protein, Optic neuropathy-inducing protein, optineurin, TFIIIA-INTP, Transcription factor IIIA-interacting protein, transcrption factor IIIA-interacting protein, tumor necrosis factor alpha-inducible cellular protein containing leucinezipper domains | |
| Recombinant protein of human Optineurin (Uniprot # Q96CV9) | |
| 100 μg | |
| Immunology, Innate Immunity, Vision | |
| 10133 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| S01-2C5 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction