missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Optineurin Polyclonal antibody specifically detects Optineurin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | Optineurin |
| Applications | Western Blot, Immunoprecipitation |
| Dilution | Western Blot, ELISA, Immunoprecipitation |
| Purification Method | Antigen and protein A Affinity-purified |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?