missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Optineurin Antibody (08), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06529UV
This item is not returnable.
View return policy
Description
Optineurin Monoclonal antibody specifically detects Optineurin in Human samples. It is validated for ELISA
Specifications
| Optineurin | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 08 | |
| ELISA | |
| FIP2FIP-2, GLC1Eglaucoma 1, open angle, E (adult-onset), HIP7ALS12, Huntingtin interacting protein L, Huntingtin yeast partner L, Huntingtin-interacting protein 7, Huntingtin-interacting protein L, HYPLHIP-7, NEMO-related protein, NRPE3-14.7K-interacting protein, Optic neuropathy-inducing protein, optineurin, TFIIIA-INTP, Transcription factor IIIA-interacting protein, transcrption factor IIIA-interacting protein, tumor necrosis factor alpha-inducible cellular protein containing leucinezipper domains | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Optineurin (NP_068815.2; Met1-Ile577). | |
| 0.1 mL | |
| Immunology, Innate Immunity, Vision | |
| 10133 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction