Learn More
Optimising solution, Elements - Mg,Al,Cr,Mn,Cu,Rh,In,Cd,Ce,Pb,Th,B,Ba for ICP 0.01mg/L, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical OP 001 500
Code : 04
Additional Details : Weight : 0.70000kg
Specifications
Optimising Solution (13 Elements) | |
Bottle | |
Optimizing Solution | |
1% HNO3 |
0.01mg/L | |
500 mL | |
Mg,Al,Cr,Mn,Cu,Rh,In,Cd,Ce,Pb,Th,B,Ba | |
Liquid |