Learn More
Optima™ LC/MS Solvents Starter Kit, Includes 1L Acetonitrile, 1L Methanol, 1L Ipa And 1L Water Optima™ LC/MS Grade
Product Code. 11988379 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 Box
Unit Size:
1 set
Product Code. Quantity Unit Size
11988379 1 Box 1 set
1 options
Product Code. 11988379 Supplier Fisher Chemical Supplier No. LCMSKIT
  • ONLINE EXCLUSIVE PRICE
    £246.00
    £230.00 / 1 sets
    Save £16.00
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
missing translation for 'validMainframeMsg'
Starter kit with reagents and amber glass bottle for development of the mobile phase for LC/MS
  • The starter kit includes a 1L amber glass bottle for each of the following reagents:
    • A955-1, Optima LC/MS Acetonitrile
    • A456-1. Optima LC/MS Methanol
    • A461-1, Optima LC/MS IPA
    • W6-1, Optima LC/MS Water
  • 1L empty amber glass bottle is included to mix reagents for the mobile phase
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Box
For Use With (Application) Creating Mobile Phase
Product Type LC/MS Solvents Starter Kit