Learn More
Grant Instruments™ Optima Bridge Plate
Description
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
- For use with Grant refrigeration units, heating and cooling circulators
- Ensures visibility/accessibility of the thermostat whilst optimising bench space
Specifications
Specifications
| For Use With (Equipment) | T Optima models to ST12, 18, 26, 38 |