missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 5 MS Accent GC Capillary Column

Brand:  Macherey-Nagel™ 725810.50

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12462820

  • £1110.00 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.2 mm
GC Capillary Column
Ideal for Ion Trap and Qadrupole MS Detectors
Gas Chromatography (GC)
0.2 μm
1 Each
G1, G2, G38
0.4 mm
Optima™ 5 MS Accent
320°C to 340°C
50 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers