missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Optifit Extended Plus Tip
GREENER_CHOICE

Product Code. 31759456 Shop All Sartorius Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 960

Product Code. 31759456

Brand: Sartorius LHX791001

Please to purchase this item. Need a web account? Register with us today!

The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware.

  • The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware
  • The tip design allows for more efficient retrieval of samples, down to the last drop
  • The longer length of these tips helps prevent contamination by ensuring that the pipette does not come into contact with the walls of labware or containers during use
  • The tips are manufactured in an ISO 8 certified cleanroom environment and are certified to be free of DNase, RNase, endotoxin and human DNA
  • Sartorius pipettes and tips are designed to seamless fit together for the most reliable and reproducible results
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 mL
Filtered Non-filtered
Sterility Sterile
Color Transparent
Autoclavable Autoclavable
Format Rack
Tip Style Ergonomic
Product Type Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.