missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Opticin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110061
This item is not returnable.
View return policy
Description
Immunogen sequence: ASLPRKERKR REEQMPREGD SFEVLPLRND VLNPDNYGEV IDLSNYEELT DYGDQLPEVK VTSLAPATSI SPAKSTTAPG TPSSNPTMTR PTTAGL.
Opticin, also known as OPTC or OPT, is a 332 amino acid protein that is secreted into the extracellular matrix and contains seven LRR repeats. Expressed in skin and fetal liver and also present in ocular tissues, such as the retina, cornea and iris, opticin functions to bind collagen fibrils and, via this interaction, is thought to regulate fibril spacing, morphology and organization. Opticin is subject to post-translational O-glycosylation and may be associated with the pathogenesis of primary open angle glaucoma.
Specifications
| Opticin | |
| Polyclonal | |
| Unconjugated | |
| OPTC | |
| dermatan sulfate proteoglycan 3; dspg3; Oculoglycan; OPT; Optc; optcl; opticin; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 269120, 304802 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.9 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q920A0 | |
| OPTC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-115 of human OPTC (NP_055174.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction