missing translation for 'onlineSavingsMsg'
Learn More

Merck Opticap™ Sterile XL 150 Express™ SHC Capsule

Product Code. 15809822 Shop All Merck Products
Click to view available options
Pore Size:
0.5/0.2 μm
This item is not returnable. View return policy

Product Code. 15809822

Brand: Merck KHGES015HB3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Autoclavable Autoclavable
Pore Size 0.5/0.2 μm
Sterility Sterile
Filtration Area 140 cm2
Outlet Style 14mm Hose barb
Type Capsule
Length (Metric) 9.7 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.