missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Optical Heat Seal, for Piko PCR plates

Product Code. 15226207
Change view
Click to view available options
Quantity:
100 sheets
Unit Size:
100 sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15226207 100 sheets 100 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 15226207 Supplier Thermo Scientific™ Supplier No. HSF0031
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

The exceptionally clear Thermo Scientific Piko PCR/qPCR Optical Heat Seals allow high transmission of fluorescence across the range of commonly-used fluorophores

When used with a Piko Thermal Cycler or PikoReal Real-Time PCR System, sample volumes as low as 5 μL can be used with minimal evaporative loss. These optical films are available in the ¼-size format for Piko PCR Plates.

Features of the Thermo Scientific Piko PCR/qPCR Film Seals
• Optical clarity with minimal auto-fluorescence for real-time PCR applications
• Tight sealing for sample volumes as low as 5 μL
• Cryo-compatible adhesive is effective to –80°C

Related Products
Optical Adhesive Film, for Piko PCR plates
TRUSTED_SUSTAINABILITY

Specifications

Color Optical
Quantity 100 sheets
For Use With (Equipment) Piko Thermal Cycler or PikoReal Real-Time PCR System
Product Type Film, Seal, or Applicator
Product Line Piko
Adhesive Optical Heat Seal
Temperature Range (Metric) -80°C
Plate Compatibility Piko Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.