missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Optical Adhesive Covers GPLE

Product Code. 17158238 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
25 covers
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17158238 25 covers Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17158238 Supplier Applied Biosystems™ Supplier No. A49767
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Applied Biosystems Optical Adhesive Cover GPLE is a pressure sensitive film that reduces well-to-well contamination and prevents sample evaporation when applied to a microplate. It is optically clear for use with real-time PCR applications.

  • A pressure sensitive film that reduces well-to-well contamination and prevents sample evaporation when applied to a microplate
  • It is optically clear for use with real-time PCR applications
  • Each lot is manufactured in an ISO registered facility, making them ideal for use with diagnostic procedures
  • Covers are easy to use, simply peel the backing away from the adhesive film and place on the microplate
  • Use in combination with the MicroAmp Adhesive Film Applicator to apply pressure and form a tight seal between the microplate and the adhesive film
  • This will reduce the possibility of cross-contamination between sample wells and prevent sample evaporation for consistent real-time PCR data
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance GPLE
Sterility Non-sterile
Includes 25 covers
Quantity 25 covers
For Use With (Application) PCR, qPCR
Content And Storage Store at room temperature and avoid direct exposure to sunlight.
Shipping Condition Room temperature
Product Type Film, Seal, or Applicator
Adhesive Pressure Sensitive
Pierceable No
Plate Compatibility 96/384-well Plate
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.