missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Opsin Green Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107203
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal. May increase spectral sensitivity in dim light. Expresssed in cone photoreceptor cells.
Specifications
| Opsin Green | |
| Polyclonal | |
| Unconjugated | |
| OPN1MW | |
| CBBM; CBD; COD5; cone dystrophy 5 (X-linked); GCP; GOP; green cone photoreceptor pigment; green cone pigment; green long wavelength sensitive cone opsin; green LWS photopigment; green-sensitive opsin; M opsin; Medium wavelength-sensitive cone opsin; medium-wave-sensitive opsin 1; Medium-wave-sensitive opsin 2; Medium-wave-sensitive opsin 3; midwavelength sensitive opsin; MWS opsin; OPN1MW; OPN1MW1; OPN1MW2; OPN1MW3; opsin 1 (cone pigments), medium-wave-sensitive; opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan); opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan), green opsin; Opsin 1 cone pigments medium-wave-sensitive 2; opsin 1 cone pigments medium-wave-sensitive 3; photopigment apoprotein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14539 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| O35599 | |
| OPN1MW | |
| Synthetic peptide located within the following region: CYLQVWLAIR AVAKQQKESE STQKAEKEVT RMVVVMVFAY CLCWGPYTFF. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction