missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Opsin 5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA585075
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%). Positive Control: HeLaS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
Specifications
| Opsin 5 | |
| Polyclonal | |
| Unconjugated | |
| OPN5 | |
| DJ402H5.1, G protein-coupled receptor 136, Gpr136, G-protein coupled receptor 136, G-protein coupled receptor PGR12, GRP136, Neuropsin, OPN5, opsin 5, opsin-5, Pgr12, TMEM13, Transmembrane protein 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221391, 353344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal, pH 7 | |
| Q6U736, Q6VZZ7 | |
| OPN5 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human neuropsin. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction