missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Opsin 1 (Medium Wave) Antibody, Alexa Fluor™ 594, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-29858AF594
This item is not returnable.
View return policy
Description
Opsin 1 (Medium Wave) Polyclonal antibody specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Opsin 1 (Medium Wave) | |
| Polyclonal | |
| Western Blot, ELISA | |
| GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2 | |
| Synthetic peptides from mouse green opsin and zebra finch red opsin. | |
| 0.1 mL | |
| GPCR, Vision | |
| 2652 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Chicken | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction