missing translation for 'onlineSavingsMsg'
Learn More

Opsin 1 (Medium Wave) Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18408267 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408267 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408267 Supplier Novus Biologicals Supplier No. NBP229858AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Opsin 1 (Medium Wave) Polyclonal antibody specifically detects Opsin 1 (Medium Wave) in Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin 1 (Medium Wave)
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias GCP, GOP, Green cone photoreceptor pigment, Green-sensitive opsin, medium-wave-sensitive opsin 1, OPN1MW, opsin 1 (cone pigments), medium-wave-sensitive 2
Host Species Chicken
Immunogen Synthetic peptides from mouse green opsin and zebra finch red opsin.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2652
Target Species Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.