missing translation for 'onlineSavingsMsg'
Learn More

Oprozomib, MedChemExpress

Product Code. 16435756
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435756

Brand: Medchem Express HY12113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oprozomib (PR-047) is an orally bioavailable and selective peptide epoxyketone proteasome inhibitor with IC50s of 36 and 82 nM for proteasome (β5) and immunoproteasome (LMP7), respectively. Oprozomib (ONX 0912) induces apoptosis in MM cells.

Oprozomib (PR-047) is an orally bioavailable and selective peptide epoxyketone proteasome inhibitor with IC50s of 36 and 82 nM for proteasome (β5) and immunoproteasome (LMP7), respectively. Oprozomib (ONX 0912) induces apoptosis in MM cells.

Specifications

Chemical Name or Material Oprozomib
Physical Form Solid
CAS 935888-69-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H32N4O7S
For Use With (Application) Cancer-Kinase/protease
Synonym ONX 0912 PR-047
Solubility Information DMSO : ≥ 50 mg/mL (93.88 mM)
SMILES C[C@]1(OC1)C([C@H](CC2=CC=CC=C2)NC([C@@H](NC([C@@H](NC(C3=CN=C(C)S3)=O)COC)=O)COC)=O)=O
Molecular Weight (g/mol) 532.61
Formula Weight 532.61
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.