Learn More
Invitrogen™ OPRM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA124628
Description
PA1-24628 is expected to cross react with bovine (93% conserved), guinea pig (100% conserved), Macaque monkey (100% conserved), and porcine (100% conserved) due to sequence homology. The molecular weight of the protein detected by PA1-24628 is 65 kDa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The OPRM1 gene encodes the mu opioid receptor, which is the primary site of action for the most commonly used opioids, including morphine, heroin, fentanyl, and methadone. It is also the primary receptor for endogenous opioid peptides beta-endorphin and the enkephalins. The OPRM1 receptor is a membrane of the G protein-coupled receptor family. There are at least 3 types of opioid receptors, mu, kappa, and delta, each with a distinct pharmacologic profile.
Specifications
| OPRM1 | |
| Polyclonal | |
| Unconjugated | |
| OPRM1 | |
| hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; OPRM; Oprm1; Oprrm1; OTTHUMP00000017463; Ror-b | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with no preservative | |
| P33535, P35372, P42866 | |
| OPRM1 | |
| NHQLENLEAETAPLP. | |
| RUO | |
| 18390, 25601, 4988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.