missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OPRM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534042
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset, Dog, Bat, Bovine, Hamster, Elephant, Panda, Pig (100%).
The OPRM1 gene encodes the mu opioid receptor, which is the primary site of action for the most commonly used opioids, including morphine, heroin, fentanyl, and methadone. It is also the primary receptor for endogenous opioid peptides beta-endorphin and the enkephalins. The OPRM1 receptor is a membrane of the G protein-coupled receptor family. There are at least 3 types of opioid receptors, mu, kappa, and delta, each with a distinct pharmacologic profile.
Specifications
| OPRM1 | |
| Polyclonal | |
| Unconjugated | |
| OPRM1 | |
| hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; OPRM; Oprm1; Oprrm1; OTTHUMP00000017463; Ror-b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100770962, 281958, 396802, 484041, 4988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P35372, P79350, Q95247 | |
| OPRM1 | |
| Synthetic 18 amino acid peptide from C-terminus of human OPRM1 / Mu Opioid Receptor. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine, Hamster, Pig, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction