Learn More
Invitrogen™ OPRD1 Recombinant Rabbit Monoclonal Antibody (HL2724)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556671
Product Code. 30276218
  • £434.00 / 100µL
Description
OPRD1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer's patients and in the putamen in Parkinson's disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OPRD1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P41143
OPRD1
Synthetic peptide encompassing a sequence within the Extracellular domain of human delta Opioid receptor. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL2724
Unconjugated
OPRD1
delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a
Rabbit
Protein A
RUO
4985
-20°C, Avoid Freeze/Thaw Cycles