missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Opipramol Di-N-Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Opipramol Di-N-Oxide
Specifications
Specifications
| Chemical Name or Material | Opipramol Di-N-Oxide |
| CAS | 2724726-55-8 |
| Analyte or Component Names | Opipramol Di-N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H29 N3 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Opipramol Di-N-Oxide |
| InChI Formula | InChI=1S/C23H29N3O3/c27-19-18-26(29)16-14-25(28,15-17-26)13-5-12-24-22-8-3-1-6-20(22)10-11-21-7-2-4-9-23(21)24/h1-4,6-11,27H,5,12-19H2 |
| SMILES | OCC[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3C=Cc4ccccc24)CC1 |
| IUPAC Name | 2-[4-(3-benzo[b][1]benzazepin-11-ylpropyl)-1,4-dioxidopiperazine-1,4-diium-1-yl]ethanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction