missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Opicapone
GREENER_CHOICE

Product Code. 18759136
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18759136

Brand: Tocris Bioscience™ 7668/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

COMT inhibitor

Opicapone is a reversible catechol O-methyltransferase inhibitor (COMT) inhibitor (IC50 = 224 nM). Decreases the ATP content and mitochondrial membrane potential in cells (IC50 values are 98 μM and 181 μM respectively). When administered with L-DOPA (Cat. No. 3788) it attenuates Parkinson's-like symptoms in an MPTP-induced animal model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[3-(2,5-Dichloro-4,6-dimethyl-1-oxido-3-pyridinyl)-1,2,4-oxadiazol-5-yl]-3-nitro-1,2-benzenediol
CAS 923287-50-7
Quantity 10 mg
Target Catechol O-Methyltransferase (COMT) Inhibitors
Molecular Formula C15H10Cl2N4O6
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.