missing translation for 'onlineSavingsMsg'
Learn More

Opicapone, MedChemExpress

Product Code. p-200098808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478837

Brand: Medchem Express HY148965mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Opicapone (BIA 9-1067) is a potent third-generation catechol-O-methyltransferase (COMT) inhibitor for the research of Parkinson's disease and motor fluctuations. Opicapone decreases the ATP content of the cells with an IC50 of 98 μM.

Opicapone (BIA 9-1067) is a potent third-generation catechol-O-methyltransferase (COMT) inhibitor for the research of Parkinson's disease and motor fluctuations. Opicapone decreases the ATP content of the cells with an IC50 of 98 μM.

Specifications

Chemical Name or Material Opicapone
CAS 923287-50-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10Cl2N4O6
Quantity 5 mg
Synonym BIA 9-1067
Solubility Information DMSO : 100 mg/mL (242.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=[N+]([O-])C(Cl)=C(C2=NOC(C3=CC([N+]([O-])=O)=C(O)C(O)=C3)=N2)C(C)=C1Cl
Molecular Weight (g/mol) 413.17
Formula Weight 413.17
Percent Purity 98.56%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.