Learn More
Julabo™ Open heating bath circulator, CORIO™ C-B13
Brand: Julabo™ 9011413
1347.43 GBP valid until 2024-05-31
Use promo code "21682" to get your promotional price.
Additional Details : Weight : 18.10000kg
Description
The Open heating bath circulators of the new CORIO™ series feature high-quality bath tanks made of stainless steel.- Model for internal applications
- Bright, white, easy-to-read display
- Very quiet
- Integrated drain screw
Working temperature range: 20°C to 100°CTemperature stability: ±0.03°CHeating capacity: 2kWCirculation capacity flow rate: 6L/minDimensions W x L x H (cm): 38 x 40 x 42Filling volume (L): 9 to 13