missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPA3 Polyclonal Antibody
OPA3 Polyclonal Antibody
Brand: Invitrogen PA545032
This item is not returnable.
View return policy
Description
OPA3 Polyclonal Antibody for Western Blot
Specifications
| OPA3 | |
| Polyclonal | |
| Unconjugated | |
| OPA3 | |
| D630048P19Rik; FLJ22187; FLJ25932; Gm1425; Gm472; MGA3; MGC75494; OPA3; OPA3 outer mitochondrial membrane lipid metabolism regulator; optic atrophy 3; optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia); Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus'); optic atrophy 3 protein; Optic atrophy 3 protein homolog; outer mitochondrial membrane lipid metabolism regulator OPA3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80207 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H6K4 | |
| OPA3 | |
| Synthetic peptide directed towards the C-terminal of human OPA3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction