missing translation for 'onlineSavingsMsg'
Learn More

OPA1 Mouse anti-Human, Mouse, Rat, Bovine, Chinese Hamster, DyLight 755, Clone: 1E8-1D9, Novus Biologicals™

Product Code. 18406433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406433 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406433 Supplier Novus Biologicals Supplier No. NBP171656IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OPA1 Monoclonal antibody specifically detects OPA1 in Human, Mouse, Rat, Bovine, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPA1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E8-1D9
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1
Host Species Mouse
Immunogen Human OPA1 [Swiss-Prot# O60313].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 4976
Target Species Human, Mouse, Rat, Bovine, Chinese Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.