missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant Medical Mask
GREENER_CHOICE

Product Code. 17700199 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Green
Honey
White
Unit Size:
Pack of 40
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17700199 Blue Pack of 40
17790189 White Pack of 40
17710209 Honey Pack of 40
17700209 Green Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17700199 Leverantör Kolmi Hopen™ Leverantörsnummer M3410130

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

These splash-resistant medical masks are designed for those with sensitive skin, providing less irritation against the face.

  • Protects environment from droplets from wearer
  • Provides protection against biological fluids
  • Effective against bacteria, particles, and viruses
  • Non-irritating inner layer
  • Inner layer is specially designed to reduce moisture and wick away sweat
  • Used by healthcare workers to reduce risk of infection or in the operating room

Specifikationer

Color Blue
Fastening Type Ties
Material Polypropylene
Style Pleated
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485
Disposable Single-use
Filter Efficiency ≥98%
For Use With (Application) Medical
Latex Free Yes
Packaging Quantity 40 Pk.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.