missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Op-Air™ One Anti-fog Medical Mask
GREENER_CHOICE

Product Code. 17720189 Shop All Medicom Products
Click to view available options
:
Op-Air One Anti-fog Medical Mask; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Style: Pleated
This item is not returnable. View return policy

Product Code. 17720189

Brand: Kolmi Hopen™ M24101F30

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Anti-fog medical masks for healthcare workers, designed to prevent glasses from fogging.

  • High filtration media (polypropylene)
  • Layers (interior to exterior): wet laid cellulose, melt blown polypropylene, spunbonded polypropylene
  • Soft, non-woven mask ties and edges
  • Anatomical folding limits exposure to face
  • Integrated anti-fog band and nose piece
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)

Specifications

Color Blue
Fastening Type Ties
Material Polypropylene
Style Pleated
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683, ISO 13485, ISO 9001
Disposable Single-use
For Use With (Application) Medical
Packaging Quantity 50 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.