Learn More
OOEP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563076
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SGNLVEITVF GRPRVQNRVK SMLLCLAWFH REHRARAEKM KHLEKNLKAH ASDPHSPQD Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 43%.
OOEP Polyclonal antibody specifically detects OOEP in Human samples. It is validated for Immunohistochemistry.Specifications
OOEP | |
Polyclonal | |
Unconjugated | |
OOEP | |
2410146L05Rik; C6orf156; Factor located in oocytes permitting embryonic development; Floped; HOEP19; KH homology domain containing 2; KH homology domain-containing protein 2; KHDC2; Moep19; OEP19; oocyte and embryo protein 19; oocyte and embryo specific protein; oocyte- and embryo-specific protein 19; oocyte expressed protein; oocyte expressed protein homolog (dog); Oocyte-expressed protein homolog; OOEP; Sddr; STAT3 downstream gene and differentiation regulator | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NGQ2 | |
OOEP | |
Recombinant protein corresponding to Human OOEP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only