missing translation for 'onlineSavingsMsg'
Learn More

Onvansertib, MedChemExpress

Product Code. p-200097219
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439397 5 mg 5mg
16426956 100 mg 100mg
16416858 10 mg 10mg
16402927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16439397 Supplier Medchem Express Supplier No. HY15828

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NMS-1286937 is a potent, selective and orally available PLK1 inhibitor, with an IC50 of 2 nM.

NMS-1286937 is a potent, selective and orally available PLK1 inhibitor, with an IC50 of 2 nM.

Specifications

Chemical Name or Material Onvansertib
CAS 1034616-18-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27F3N8O3
Quantity 5 mg
Synonym NMS-1286937 NMS-P937
Solubility Information DMSO : 21 mg/mL (39.44 mM; Need ultrasonic and warming)
SMILES O=C(C1=NN(CCO)C2=C1CCC3=CN=C(NC4=CC(N5CCN(C)CC5)=CC=C4OC(F)(F)F)N=C23)N
Molecular Weight (g/mol) 532.52
Formula Weight 532.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.