missing translation for 'onlineSavingsMsg'
Learn More

Ononin, MedChemExpress

Product Code. 16479036
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479036 100 mg 100mg
16448928 10 mg 10mg
16404807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16479036 Supplier Medchem Express Supplier No. HYN0270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ononin is an isoflavonoid, is an additional growth inhibitor in soils associated with the weed, Pluchea lanceolata.

Ononin is an isoflavonoid, is an additional growth inhibitor in soils associated with the weed, Pluchea lanceolata.

Specifications

Chemical Name or Material Ononin
CAS 486-62-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22O9
Quantity 100 mg
Synonym Ononoside Formononetin 7-O-β-D-glucopyranoside
Solubility Information DMSO : 100 mg/mL (232.34 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)[C@H](O)[C@@H](CO)O1)[C@@H]1OC2=CC=C(C(C(C3=CC=C(OC)C=C3)=CO4)=O)C4=C2
Molecular Weight (g/mol) 430.4
Formula Weight 430.4
Percent Purity 99.86%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.