missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ononetin
GREENER_CHOICE

Product Code. 30001986
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30001986

Brand: Tocris Bioscience™ 5143/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRPM3 blocker

Naturally occurring deoxybenzoin. TRPM3 channel blocker (IC50 = 300 nM). Inhibits pregnenolone sulfate-induced intracellular Ca2+ accumulation in HEKmTRPM3 cells and dorsal root ganglia (DRG) neurons in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 487-49-0
Quantity 50 mg
Molecular Formula C15H14O4
Formula Weight 258.27
Molecular Weight (g/mol) 258.273
Chemical Name or Material Ononetin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.