missing translation for 'onlineSavingsMsg'
Learn More

ONO-AE3-208, MedChemExpress

Product Code. 16410598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16410598

Brand: Medchem Express HY50901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ONO-AE3-208 is a selective and orally active EP4 receptor antagonist with a Ki of 1.3 nM. ONO-AE3-208 shows less potently affects EP3, FP, and TP receptors (Ki of 30 nM, 790 nM, and 2400 nM, respectively). ONO-AE3-208 suppresses cell invasion, migration, and metastasis of prostate cancer.

ONO-AE3-208 is a selective and orally active EP4 receptor antagonist with a Ki of 1.3 nM. ONO-AE3-208 shows less potently affects EP3, FP, and TP receptors (Ki of 30 nM, 790 nM, and 2400 nM, respectively). ONO-AE3-208 suppresses cell invasion, migration, and metastasis of prostate cancer.

Specifications

Chemical Name or Material ONO-AE3-208
CAS 402473-54-5
Color White
Purity Grade Notes Research
Molecular Formula C24H21FN2O3
Quantity 5 mg
Synonym AE 3-208
Solubility Information DMSO : 33.33 mg/mL (82.41 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C(NC(C(C2=C3C=CC=CC3=C(F)C=C2)C)=O)=C1)CCCC(O)=O
Molecular Weight (g/mol) 404.43
Formula Weight 404.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.