missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ongericimab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Ongericimab Humanized |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30284172
![]() |
Invitrogen™
MA559617 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284158
|
Invitrogen™
MA559618 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ongericimab, also known as a recombinant humanized anti-PCSK9 monoclonal antibody, is primarily used to treat hypercholesterolemia by reducing low-density lipoprotein cholesterol (LDL-C) levels. The PCSK9 gene, located on chromosome 1p32.3, spans 22 kilobases and consists of 12 exons and 11 introns. It encodes a 692 amino acid glycoprotein involved in lipid metabolism, comprising a signal peptide (amino acids 1-30), a prosegment or inhibitory prodomain (amino acids 31-152), a subtilisin-like catalytic domain (amino acids 153-452), and a cysteine-histidine-rich C-terminal domain. PCSK9 plays a pivotal role in cholesterol homeostasis by promoting the degradation of low-density lipoprotein receptors (LDLR) in the liver, thereby increasing plasma LDL levels. Mutations in PCSK9 can lead to disorders such as familial hypercholesterolemia. In addition to its well-established role in cardiovascular diseases, emerging research indicates that PCSK9 variants may influence cancer outcomes and that PCSK9 inhibition can enhance the efficacy of immune checkpoint therapies.Specifications
| Ongericimab Humanized | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG4SP | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title