missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Red, 555

Product Code. 18303273 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 Assays
Unit Size:
20 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18303273 20 Assays 20 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18303273 Supplier Novus Biologicals™ Supplier No. NBP31209320ASSAYS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for in situ apoptosis detection of tissue samples (Paraffin embedding, freezing and ultrathin section) and cells (Cell smears, cell climbing smears).

TRUSTED_SUSTAINABILITY

Specifications

Description OneStep TUNEL
Content And Storage Store at -20C.
Quantity 20 Assays
For Use With (Application) Flow Cytometry, Fluorescence Imaging
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.