missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ OneFeed™ Supplement

Product Code. p-7084485
Click to view available options
Quantity:
2 L
10 L
50 L
Unit Size:
10 L
2 L
50 L
This item is not returnable. View return policy

Product Code. 16515611

Brand: Gibco™ 670110

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A versatile medium supplement.

Gibco™ OneFeed™ Supplement is a chemically-defined, animal-free, protein-free feed shown to enhance protein production when used in a variety of commercially available medium.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO DG44, BHK-21, CHO-K1, CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Format Container
Quantity 2 L
Source Animal origin-free
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.