missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ONECUT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119822
This item is not returnable.
View return policy
Description
Positive Control: SW480 cell lysate, Hela cell lysate, human stomach tissue lysate, AGS cell lysate, human colon carcinoma tissue, human stomach tissue, SW620. Subcellular Location: Nucleus.
Transcriptional activator. Binds the consensus DNA sequence 5'-DHWATTGAYTWWD-3' on a variety of gene promoters such as those of HNF3B and TTR.
Specifications
| ONECUT3 | |
| Polyclonal | |
| Unconjugated | |
| ONECUT3 | |
| OC3; OC-3; One cut domain family member 3; one cut domain, family member 3; one cut homeobox 3; Onecut3; Transcription factor ONECUT-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 390874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O60422 | |
| ONECUT3 | |
| Synthetic peptide within human ONECUT3 aa 430-470. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction