missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ONECUT3 Antibody (10B2F5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37340-0.025ml
This item is not returnable.
View return policy
Description
ONECUT3 Monoclonal specifically detects ONECUT3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ONECUT3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| OC3, OC-3, one cut domain family member 3, one cut domain, family member 3, one cut homeobox 3, transcription factor ONECUT-3 | |
| Mouse | |
| 50 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 10B2F5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ONECUT3 | |
| Purified recombinant fragment of human ONECUT3 (AA: 350-455) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 390874 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction