Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
Brand:  Hamilton™ 90662
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11593134
  • £41.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
  • Fitting manufactured from CTFE
  • Tubing manufactured from PTFE
  • For female luers
  • A variety of gauges and lengths available
Specifications
Specifications
CTFE (Fitting), PTFE (Tubing)
Custom
1/Pk.
Female Luer
12 G
Tubing Assembly
12 ga. Connections