Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
Brand: Hamilton™ 90676
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Fitting manufactured from CTFE
- Tubing manufactured from PTFE
- For female luers
- A variety of gauges and lengths available
Specifications
CTFE (Fitting), PTFE (Tubing) | |
Custom | |
1/Pk. | |
Female Luer |
26 G | |
Tubing Assembly | |
26 ga. Connections |