missing translation for 'onlineSavingsMsg'
Learn More

Ondansetron, MedChemExpress

Product Code. 16408056
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408056 100 mg 100mg
16498318 10 mg 10mg
16414207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16408056 Supplier Medchem Express Supplier No. HYB0002B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ondansetron(GR 38032; SN 307) is a serotonin 5-HT3 receptor antagonist used mainly as anantiemetic (to treat nausea and vomiting), often following chemotherapy.

Ondansetron(GR 38032

Specifications

Chemical Name or Material Ondansetron
CAS 99614-02-5
Color White
Purity Grade Notes Research
Molecular Formula C18H19N3O
Quantity 100 mg
Synonym GR 38032 SN 307
Solubility Information DMSO : 10 mg/mL (34.09 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(CN2C=CN=C2C)CCC(N3C)=C1C4=C3C=CC=C4
Molecular Weight (g/mol) 293.36
Formula Weight 293.36
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.