missing translation for 'onlineSavingsMsg'
Learn More

Oncrasin-1, MedChemExpress

Product Code. p-200097613
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16457108

Brand: Medchem Express HY16662

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oncrasin-1 is a potent and effective anticancer inhibitor that kills various human lung cancer cells with K-Ras mutations at low or submicromolar concentrations; also led to abnormal aggregation of PKCι in nucleus of sensitive cells but not in resistant cells.

Oncrasin-1 is a potent and effective anticancer inhibitor that kills various human lung cancer cells with K-Ras mutations at low or submicromolar concentrations

Specifications

Chemical Name or Material Oncrasin-1
CAS 75629-57-1
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C16H12ClNO
Quantity 10 mg
Solubility Information DMSO : ≥ 43 mg/mL (159.42 mM)
SMILES O=CC1=CN(CC(C=C2)=CC=C2Cl)C3=CC=CC=C31
Molecular Weight (g/mol) 269.73
Formula Weight 269.73
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.