missing translation for 'onlineSavingsMsg'
Learn More

Oncrasin-1, MedChemExpress

Product Code. 16457108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457108 10 mg 10mg
16443147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16457108 Supplier Medchem Express Supplier No. HY16662
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oncrasin-1 is a potent and effective anticancer inhibitor that kills various human lung cancer cells with K-Ras mutations at low or submicromolar concentrations; also led to abnormal aggregation of PKCι in nucleus of sensitive cells but not in resistant cells.

Oncrasin-1 is a potent and effective anticancer inhibitor that kills various human lung cancer cells with K-Ras mutations at low or submicromolar concentrations

Specifications

Chemical Name or Material Oncrasin-1
CAS 75629-57-1
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C16H12ClNO
Quantity 10 mg
Solubility Information DMSO : ≥ 43 mg/mL (159.42 mM)
SMILES O=CC1=CN(CC(C=C2)=CC=C2Cl)C3=CC=CC=C31
Molecular Weight (g/mol) 269.73
Formula Weight 269.73
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.