missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (17001.31), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10842
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize | |
| OSM | |
| Recombinant full length protein (Human) expressed in E. coli. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 5008 | |
| To one vial of lyophilized powder, add 1 ml of 0.2 um-filtered PBS to produce a 500 ug/ml stock solution. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Blocking Assay | |
| 17001.31 | |
| Unconjugated | |
| MGC20461, oncostatin M, oncostatin-M | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Does not cross-react with recombinant human IL-6, IL-11, CNTF, LIF and recombinant mouse OSM. | |
| Human, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction