missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (17001.31), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10842R
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| OSM | |
| Recombinant full length protein (Human) expressed in E. coli. | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5008 | |
| Human, Mouse (Negative) | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 17001.31 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| MGC20461, oncostatin M, oncostatin-M | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Does not cross-react with recombinant human IL-6, IL-11, CNTF, LIF and recombinant mouse OSM. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction