missing translation for 'onlineSavingsMsg'
Learn More

Oncostatin M/OSM Antibody (015), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18397050 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397050 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397050 Supplier Novus Biologicals Supplier No. NBP289506AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oncostatin M/OSM
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 15
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias MGC20461, oncostatin M, oncostatin-M
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Oncostatin M/OSM (P13725; Met1-Arg221).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5008
Test Specificity No cross-reactivity in ELISA with: Mouse OSM Human CNTF Human IL2 Human IL6 Human LIF Human GCSF
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.