missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nepuvibart Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA562774
This item is not returnable.
View return policy
Description
Nepuvibart Recombinant Monoclonal Antibody for Flow, ELISA, Functional Assay
Nepuvibart targets GM-CSF implicated in inflammatory disorders like rheumatoid arthritis and COVID-19.
Specifications
| Nepuvibart | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CAS:2640224-48-0; ZRC-3308; ZRC-3308B10 | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Functional Assay | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction